Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Connectors, with GL thread

Connectors, with GL thread

6,5 mm, GL 14
Katalognr. 229-0000
Anbieter:   Bohlender
Connectors, with GL thread
Katalognr. 229-0000
Anbieter:  Bohlender
Connectors, with GL thread
Connectors, with GL thread

6,5 mm, GL 14

Preis
SFr. 44.20
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents

PTFE

  • Very good chemical resistance, the fluid medium only comes into contact with PTFE
  • Temperature resistant from −200 to +250 °C
  • Pressure resistant up to a maximum of 10 bar, vacuum suitable

Connector with two, three or four GL screw connections. To distribute liquids or gases. Can be used as a reducer to connect different tubes or hoses.

Technische Daten
  • Hole Ø:
    6,5 mm
  • Thread:
    GL 14
  • Seq:
    01