ContactUsLinkComponent
Thermometer, precision, mercury filled, DIN certified, measuring range:-20:50/0.1C, enclosed scale, capllary lens front, colourles s , Richter-top finish with button, dimension:450x9-9.5mm, suitable for government verification. Until 10th October 2 1 * 1 items
Catalog # AMARL25996
Supplier:  Amarell
Thermometer, precision, mercury filled, DIN certified, measuring range:-20:50/0.1C, enclosed scale, capllary lens front, colourles s , Richter-top finish with button, dimension:450x9-9.5mm, suitable for government verification. Until 10th October 2 1 * 1 items
Catalog # AMARL25996
Supplier:  Amarell
Frequently Bought Together