ContactUsLinkComponent
pH combination electrode with plug head L32. Plastic shaft, fibre-diaphragm, electrolyte KCl 3 mol/l, Silamid-reference system, sph ere membrane, A-glass, length 120 mm, 12 mm , -5...+50 C, 1...13 pH 1 * 1 items
Catalog # SCOI1061093
Supplier:  SI Analytics
pH combination electrode with plug head L32. Plastic shaft, fibre-diaphragm, electrolyte KCl 3 mol/l, Silamid-reference system, sph ere membrane, A-glass, length 120 mm, 12 mm , -5...+50 C, 1...13 pH 1 * 1 items
Catalog # SCOI1061093
Supplier:  SI Analytics
Frequently Bought Together