ContactUsLinkComponent
Flask volumetric, capacity 5ml, class A, PE Stoppered, tolerance 0.04ml, height 70mm, NS-ground 10/19, graduation color blue, conf o rmity certified. 1 * 2 items
Catalog # HIRS2820153_S
Supplier:  HIRSCHMANN
undefined
Flask volumetric, capacity 5ml, class A, PE Stoppered, tolerance 0.04ml, height 70mm, NS-ground 10/19, graduation color blue, conf o rmity certified. 1 * 2 items
Catalog # HIRS2820153_S
Supplier:  HIRSCHMANN
Frequently Bought Together