Pipette volumetric, capacity 2.5ml, class AS, amber graduation, soda-lime glass, tolerance 0.01ml, total length +-10mm:350mm, confor mity certified. 1 * 1 items
Catalog # HIRS1340145_S
Supplier: HIRSCHMANN
Pipette volumetric, capacity 2.5ml, class AS, amber graduation, soda-lime glass, tolerance 0.01ml, total length +-10mm:350mm, confor mity certified. 1 * 1 items
Catalog # HIRS1340145_S
Supplier: HIRSCHMANN
Frequently Bought Together
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection