ContactUsLinkComponent
pH-combination electrode A 164 with plug head, glass shaft, ground joint-diaphragm, electrolyte KCl 3 mol/l, Silamid reference, tem perature sensor Pt 1000, sphere membrane, A glass, SMEK plug head, length 170 mm, 12 mm , -5...+100 C, 0...14 pH 1 * 1 items
Catalog # 662-1970
Supplier:  SI Analytics
pH-combination electrode A 164 with plug head, glass shaft, ground joint-diaphragm, electrolyte KCl 3 mol/l, Silamid reference, tem perature sensor Pt 1000, sphere membrane, A glass, SMEK plug head, length 170 mm, 12 mm , -5...+100 C, 0...14 pH 1 * 1 items
Catalog # 662-1970
Supplier:  SI Analytics
Frequently Bought Together