Pipette tips
Bevelled, non-graduated, 200 - 1000 µl, -, 76 mm, Blue, 96 tips/rack; 6 racks/case
Catalog # ALPCLW6470
Supplier: Alpha Laboratories
Jump to:
Product Details & Documents
These tips are manufactured from medical grade PP in modern production facilities with stringent quality controls at every stage.
- Tips are lot tested and certified for absence of detectable human DNA and detectable endotoxins (pyrogens)
- Tips are tested and certified for absence of heavy metals, detectable DNase and RNase
- With graduations to visually check volume
- Suitable for all major pipettor brands
Specifications
- Colour code:Blue
- Packed:96 tips/rack; 6 racks/case
- Sterile:-
- Volume:200 - 1000 µl
- Length:76 mm
- Tip style:Bevelled, non-graduated