Orders LinkContactUsLinkComponent
Face shields, Bionic™
Uncoated, Acetate
  111-0252
Face shields, Bionic™
  111-0252
Face shields, Bionic™
Face shields, Bionic™
Uncoated, Acetate
67.90 SFr.
1 * 1 ST
This innovative face shield provides protection from extreme mechanical hazards. It can be worn in combination with safety spectacles or respiratory protection.
  • With extended protection for brow and chin
  • Locking mechanism for added safety
  • Extra soft padding with washable or replaceable sweatband
  • Various adjustments for best comfort
Weight: 355 g.
EN 166, EN 169, EN 170, 1 F, B, T, C, 3, 8, 9
 : EN 166, EN 169, EN 170, 1 F, B, T, C, 3, 8, 9
  • Type:
    Uncoated
  • Material:
    Acetate
Accessories
  •  
    Description
    Material
  • 111-0740
    Replacement visor, uncoated
    PC
    1 * 1 ST - SFr. 16.00
  •  
    Description
    Material
  • 111-2160
    Replacement visor, uncoated
    Acetate
    1 * 1 ST - SFr. 26.10
  •  
    Description
    Material
  • 111-1293
    Replacement visor, anti-scratch/anti-fog
    PC
    1 * 1 ST - SFr. 41.30
  •  
    Description
  • 113-8053
    Replacement sweatband
    1 * 1 ST - SFr. 9.40