Vials with screw thread, Nalgene®
Serum vial, 10,00 ml, Amber, No, 20-415, +
Catalog # 215-1505
Supplier: Thermo Fisher Scientific
Vials with screw thread, Nalgene®
Catalog # 215-1505
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
These vials are ideal for diagnostic and biological sera, pharmaceuticals for veterinary and ophthalmologic usage as well as many other applications. They are made from clear or coloured brown PETG.
- Available clear or coloured brown, sterile or non-sterile
- A wide range of different screw caps is optionally available
- Manufactured and packed in workrooms equipped with HEPA filters
Closures for vials: HDPE, coloured. For safe and tight closure of narrow-neck vials. Available in non-sterile or sterile versions. Non sterile caps are loose packed, sterile caps are loose packed in a box with a three-section insert.
For safe and tight closure of narrow-neck vials.
Documents
Certificates
Search by Lot NumberSpecifications
- Cap included:No
- Cap Size:20-415
- Sterile:+
- Capacity:10,00 ml
- Colour:Amber
- Description:Serum vial
- Cat. No.:215-1505
- Environmental certification:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Low Manufacturing Impact



