Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Tissue grinders, Potter type

Tissue grinders, Potter type

15 ml, 16×155 mm
Catalog # KART6115
Supplier:   KARTELL
Tissue grinders, Potter type
Catalog # KART6115
Supplier:  KARTELL
Tissue grinders, Potter type
Tissue grinders, Potter type

15 ml, 16×155 mm

Price
SFr. 145.00
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Int.Ø×H:
    16×155 mm
  • Capacity:
    15 ml
  • Seq:
    04