Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Scintillation vials

Scintillation vials

20,00 ml, 26,5×58,5 mm, Yes, Screw cap
Catalog # DELT900100
Supplier:   DELTALAB
Scintillation vials
Catalog # DELT900100
Supplier:  DELTALAB
Scintillation vials

20,00 ml, 26,5×58,5 mm, Yes, Screw cap

Price
SFr. 456.00
Log in for availability
1 * 1.000 ST
Product Details & Documents

Single use vials made of high density polyethylene to minimise solvent losses.

  • Screw cap ensures leakproof handling
  • Compatible with most liquid scintillation counters

Documents

Specifications
  • Cap included:
    Yes
  • Ø×H:
    26,5×58,5 mm
  • Capacity:
    20,00 ml
  • Seq:
    02
  • Cap type:
    Screw cap
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.