Glass beads
Glass beads, solid, 1000 g pack, 1 mm
Catalog # MARI4901001
Supplier: MARIENFELD
Jump to:
Product Details & Documents
Made of soda lime glass.
- Solid
- Polished
Glass beads are used for mixing or milling. The smaller the beads and the higher their quantity the more intense is the mixing or milling.
Packaging: In poly bag of 500 g or 1000 g each.
Documents
Specifications
- Ø:1 mm
- Description:Glass beads, solid, 1000 g pack
- Seq:01
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



