Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Glass beads

Glass beads

Glass beads, solid, 1000 g pack, 6 mm
Catalog # MARI4901006
Supplier:   MARIENFELD
Glass beads
Catalog # MARI4901006
Supplier:  MARIENFELD
Glass beads
Glass beads

Glass beads, solid, 1000 g pack, 6 mm

Price
SFr. 47.30
Log in for availability
1 * 1 kg
Product Details & Documents

Made of soda lime glass.

  • Solid
  • Polished

Glass beads are used for mixing or milling. The smaller the beads and the higher their quantity the more intense is the mixing or milling.

 : In poly bag of 500 g or 1000 g each.

Documents

Specifications
  • Ø:
    6 mm
  • Description:
    Glass beads, solid, 1000 g pack
  • Seq:
    06