Orders LinkContactUsLinkComponent
Adapters for GL screw threads
Adapters for GL screw threads
Adapters with conical ground joint 14/23, GL 18
Catalog # QVFLHAA6012504
Supplier:   QVF LABORTECHNIK
Adapters for GL screw threads
Catalog # QVFLHAA6012504
Supplier:  QVF LABORTECHNIK
Adapters for GL screw threads
Adapters with conical ground joint 14/23, GL 18
Price
Inquire For Price
Log in for availability
1 * 10 ST
Product Details & Documents
For connecting GL screw threads and conical ground joints.
Specifications
  • Thread:
    GL 18
  • Description:
    Adapters with conical ground joint 14/23
  • Seq:
    01