Solid phase extraction cartridges, Bond Elut™ PBA
Straight barrel cartridge, PBA, 60 Å, 40 µm, 3 ml, 500 mg
VARI12102105
: VARIAN
PBA is a unique sorbent consisting of a phenylboronic acid covalently linked to a silica gel surface. Covalent processes result in a higher retention to the sorbent surface than other mechanisms, resulting in superior clean-up. The boronate group has a high specificity for cis-diol containing compounds such as catechols, nucleic acids, low molecular weight proteins and carbohydrates.
- High specificity for cis-diol compounds
- Very effective in separating DNA and RNA
Bond Elut™ PBA can also be used to retain oligomeric materials such as polyethylene glycols (PEGs) from a variety of aqueous or biological matrices.
- Particle Size:40 µm
- Pore size:60 Å
- Capacity:3 ml
- Description:Straight barrel cartridge
- Seq:04
- Bed weight:500 mg
- Phase:PBA
- Carbon load:7,9%
- Cat. No.:VARI12102105
- Endcapped:No
- Surface area:500 m²/g



