Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ Focus

Solid phase extraction cartridges, Bond Elut™ Focus

Straight barrel cartridge, Focus, 120 Å, 44 µm, 6 ml, 50 mg
Catalog # VARIA5606050
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ Focus
Catalog # VARIA5606050
Supplier:  VARIAN
Solid phase extraction cartridges, Bond Elut™ Focus

Straight barrel cartridge, Focus, 120 Å, 44 µm, 6 ml, 50 mg

Price
Inquire For Price
Log in for availability
1 * 100 ST
Product Details & Documents

Bond Elut™ Focus solid phase extraction cartridges with enhanced polar recovery. Features a unique amide functionalised polar-enhanced sorbent technology that delivers outstanding retention for polar and non-polar analytes.

  • Simple, universal method reduces method validation times
  • 'Power Rinse' delivers cleaner extracts, improving sensitivity
  • Hydrogen bond donor capability increases retention of basic analytes

Rigorous washes using 10 to 20% ACN and MeOH can be used to aggressively remove endogenous matrix interferences. For very polar analytes that are held by strictly polar interactions.

Specifications
  • Particle Size:
    44 µm
  • Pore size:
    120 Å
  • Capacity:
    6 ml
  • Description:
    Straight barrel cartridge
  • Bed weight:
    50 mg
  • Phase:
    Focus
Accessories

Product Information

  • Catalog #
    Filling
    Availability
    Price
  • 96-round well plate for SPE, Focus™
    96-round well plate for SPE, Focus™
    VARIA59660
    10 mg
    Log in for availability
    1 * 1 ST