Orders LinkContactUsLinkComponent
Insulin, Reference Standards
Insulin, Reference Standards
Insulin Pork, USP reference standard, Vial, 50 mg
Catalog # 1342300.
Supplier:   USP
Insulin, Reference Standards
Catalog # 1342300.
Supplier:  USP
CAS Number:  
Insulin, Reference Standards
Insulin Pork, USP reference standard, Vial, 50 mg
CAS Number:  
Price
524.00 SFr.
Log in for availability
1 * 50 mg
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Insulin Glargine for Peak Identication: Mixture of Insulin Glargine and 0A-Arg-Insulin Glargine
Certifications: US Pharmacopea Reference Standard
Caution: Cold Shipment required
Documents
Specifications
Specifications
  • Pack type:
    Vial
  • Pk:
    50 mg
  • Standards:
    USP reference standard
  • Description:
    Insulin Pork
  • Cat. No.:
    1342300.
  • Description:
    Insulin Pork
  • CAS No.:
    12584-58-6