Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Stirring rods

Stirring rods, glass, with fused ends, Glass, 3 mm, 200 mm
Catalog # 441-0074
Stirring rods
Catalog # 441-0074
Stirring rods
Stirring rods

Stirring rods, glass, with fused ends, Glass, 3 mm, 200 mm

Price
35.90 SFr.
Log in for availability
1 * 100 ST
Product Details & Documents

AR-Glas®. With fused ends.

Specifications
  • Ø:
    3 mm
  • Description:
    Stirring rods, glass, with fused ends
  • Length:
    200 mm
  • Material:
    Glass
  • Seq:
    014