Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Container, with screw cap

Container, with screw cap

500 ml, 104 mm, 100 mm
Catalog # HUEN915300
Supplier:   hünersdorff GmbH
Container, with screw cap
Catalog # HUEN915300
Supplier:  hünersdorff GmbH
Container, with screw cap
Container, with screw cap

500 ml, 104 mm, 100 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

Containers with screw cap round, for varied applications, made of food-safe natural polypropylene.

  • Resistant to acids and chemicals
  • With tightly closing lid
  • Deep freezable
Specifications
  • Ext.Ø:
    104 mm
  • Capacity:
    500 ml
  • Height:
    100 mm
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.