Curcumin
Glass bottle for solids, 5 g
Catalog # CAYM81025-5
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
Synonyms: (1E,6E)-1,7-Bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione
- CAS Number:458-37-7
- Formula:C₂₁H₂₀O₆
- MDL Number:MFCD00008365
- Molecular Weight:368.39 g/mol
- EINECS:207-280-5
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



