Orders LinkContactUsLinkComponent
Forceps
Forceps
Straight, short fine tips, 110 mm
Catalog # 241-0331
Supplier:   EXCELTA
Forceps
Catalog # 241-0331
Supplier:  EXCELTA
Forceps
Straight, short fine tips, 110 mm
Price
74.50 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel, antimagnetic.
  • Economical special forceps for precision applications
  • Corrosion-resistant
Ideal when working with microelectronic assemblies and in the electronics industry.
Specifications
  • Description:
    Straight, short fine tips
  • Length:
    110 mm
  • Seq:
    01