Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Microsette™ M503

Biopsy processing/embedding cassettes, Microsette™ M503

Blue
Catalog # 720-0965
Supplier:   Simport Scientific
Biopsy processing/embedding cassettes, Microsette™ M503
Catalog # 720-0965
Supplier:  Simport Scientific
Biopsy processing/embedding cassettes, Microsette™ M503
Biopsy processing/embedding cassettes, Microsette™ M503

Blue

Price
SFr. 623.00
Log in for availability
1 * 1.000 ST
Product Details & Documents

Acetal polymer.

  • No need for biopsy pads
  • Six compartments, each 7×12 mm, numbered 1 to 6
  • 2000 square openings (each 0,26 mm) to maximise fluid exchange
  • Integral lid
  • Anterior writing area 45° angle suitable for some labelling systems
Specifications
  • Colour:
    Blue