Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Biopsy processing/embedding cassettes, Microsette™ M503

White
  720-0961
Biopsy processing/embedding cassettes, Microsette™ M503
  720-0961
Biopsy processing/embedding cassettes, Microsette™ M503
Biopsy processing/embedding cassettes, Microsette™ M503

White

 
SFr. 623.00
   
1 * 1.000 ST
 

Acetal polymer.

  • No need for biopsy pads
  • Six compartments, each 7×12 mm, numbered 1 to 6
  • 2000 square openings (each 0,26 mm) to maximise fluid exchange
  • Integral lid
  • Anterior writing area 45° angle suitable for some labelling systems
 
  • Colour:
    White