Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Microsette™ I M502

Biopsy processing/embedding cassettes, Microsette™ I M502

White
Catalog # 720-0951
Supplier:   Simport Scientific
Biopsy processing/embedding cassettes, Microsette™ I M502
Catalog # 720-0951
Supplier:  Simport Scientific
Biopsy processing/embedding cassettes, Microsette™ I M502
Biopsy processing/embedding cassettes, Microsette™ I M502

White

Price
SFr. 623.00
Log in for availability
1 * 1.000 ST
Product Details & Documents

Acetal polymer.

  • No need for biopsy pads
  • Large compartment 25×30 mm
  • 2000 holes (Ø 0,26 mm) suitable for needle biopsies
  • Integral lid
  • Anterior writing area 45° angle suitable for some labelling systems
Specifications
  • Colour:
    White