Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Microsette™ I M502

Biopsy processing/embedding cassettes, Microsette™ I M502

Purple
Catalog # 720-0949
Supplier:   Simport Scientific
Biopsy processing/embedding cassettes, Microsette™ I M502
Catalog # 720-0949
Supplier:  Simport Scientific
Biopsy processing/embedding cassettes, Microsette™ I M502
Biopsy processing/embedding cassettes, Microsette™ I M502

Purple

Price
SFr. 623.00
Log in for availability
1 * 1.000 ST
Product Details & Documents

Acetal polymer.

  • No need for biopsy pads
  • Large compartment 25×30 mm
  • 2000 holes (Ø 0,26 mm) suitable for needle biopsies
  • Integral lid
  • Anterior writing area 45° angle suitable for some labelling systems
Specifications
  • Colour:
    Purple
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.