Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Biopsy processing/embedding cassettes, Micromesh™ M508

Blue
  720-0986
Biopsy processing/embedding cassettes, Micromesh™ M508
  720-0986
Biopsy processing/embedding cassettes, Micromesh™ M508
Biopsy processing/embedding cassettes, Micromesh™ M508

Blue

 
SFr. 616.00
   
1 * 1.000 ST
 

Acetal polymer.

  • No need for biopsy pads
  • Cassettes have four square compartments each 13 mm
  • Cover and base have about 1676 square openings to maximise fluid exchange
  • Integral lid
  • Anterior writing area 45° angle suitable for some labelling systems
 
  • Colour:
    Blue