Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Micromesh™ M508

Biopsy processing/embedding cassettes, Micromesh™ M508

Pink
Catalog # 720-0983
Supplier:   Simport Scientific
Biopsy processing/embedding cassettes, Micromesh™ M508
Catalog # 720-0983
Supplier:  Simport Scientific
Biopsy processing/embedding cassettes, Micromesh™ M508
Biopsy processing/embedding cassettes, Micromesh™ M508

Pink

Price
SFr. 616.00
Log in for availability
1 * 1.000 ST
Product Details & Documents

Acetal polymer.

  • No need for biopsy pads
  • Cassettes have four square compartments each 13 mm
  • Cover and base have about 1676 square openings to maximise fluid exchange
  • Integral lid
  • Anterior writing area 45° angle suitable for some labelling systems
Specifications
  • Colour:
    Pink
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.