Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Hand towels, SCOTT®

SCOTT®, single sheet roll, 1 ply, Blue, 304 m × 200 mm, 6 rolls/carton
Catalog # KIMW6668
Supplier:   KIMBERLY CLARK
Hand towels, SCOTT®
Catalog # KIMW6668
Supplier:  KIMBERLY CLARK
Hand towels, SCOTT®
Hand towels, SCOTT®

SCOTT®, single sheet roll, 1 ply, Blue, 304 m × 200 mm, 6 rolls/carton

Price
SFr. 241.00
Log in for availability
1 * 6 ROU
Product Details & Documents

Disposable hand towels made from AIRFLEX* (a voluminous, textile-like fabric).

  • High absorption performance, and tear-proof even when wet
  • Single, self-presenting towels for enhanced hygiene and less waste
  • Extremely soft
Specifications
  • Packed:
    6 rolls/carton
  • Colour:
    Blue
  • Description:
    SCOTT®, single sheet roll, 1 ply
  • L×W:
    304 m × 200 mm
  • Seq:
    06
Accessories

Product Information

  • Catalog #
    W×D×H
    Availability
    Price
  • Dispenser for hand towel rolls, AQUARIUS* 6959, white ABS, with window
    Dispenser for hand towel rolls, AQUARIUS* 6959, white ABS, with window
    115-1779
    326×241×430 mm
    Log in for availability
    1 * 1 ST - SFr. 245.00