Order Entry
Switzerland
Orders LinkContactUsLinkComponent
pH/°C testers, 206

pH/°C testers, 206

pH/°C tester, 206 pH2, single instrument
Catalog # TEST0563.2062
Supplier:   Testo
pH/°C testers, 206
Catalog # TEST0563.2062
Supplier:  Testo
pH/°C testers, 206
pH/°C testers, 206

pH/°C tester, 206 pH2, single instrument

Price
SFr. 308.00
Log in for availability
1 * 1 ST
Product Details & Documents

The testo 206 series is ideal for pH measurements in liquid and semi-solid media. The instruments offer a combination of a pH immersion probe and a temperature probe for precise and fast temperature compensation. Thanks to the large volume of gel electrolyte and the dual wall diaphragm, the probe is leak-proof, maintenance-free and insensitive to dirt.

  • Built-in temperature sensor
  • Maintenance-free gel electrolyte
  • Robust, waterproof and dishwasher-safe protective cover (TopSafe, protection class IP 65)
  • Automatic final value recognition (Auto-hold)
  • 1, 2, or 3-point calibration possible

Testo 206-pH1 is specially designed for fast pH checks in liquids.

Testo 206-pH2 is ideally suited for pH measurement in semi-solid and protein-rich foods such as jellies, creams, cheese, fruit etc.

Testo 206-pH3 is equipped with a BNC socket which allows the connection of any pH probes to the instrument, depending on the application. This makes the instrument universally applicable.

Delivery information: Single instruments are supplied with TopSafe cover and belt/wall holder. pH1 and pH2 also include a storage cap with gel.
pH1 and pH2 starter kits are supplied with storage cap with gel, calibration dosing bottles 250 ml pH 4 and 7, TopSafe cover, belt/wall holder and aluminium case.
All instruments are supplied with probe head but without probe. Probes need to be ordered separately.

Specifications
  • Description:
    pH/°C tester, 206 pH2, single instrument
  • Compliance:
    IP68
  • pH range:
    0...+14
  • Power supply:
    1 × CR2032 battery
  • Temperature compensation:
    Automatic
  • pH calibration:
    1, 2, or 3-point calibration
  • Temperature resolution:
    0,1 °C
  • Temperature accuracy:
    ±0,4 °C
  • Weight:
    0,062 kg
  • Cat. No.:
    TEST0563.2062
  • pH accuracy:
    ±0,02
  • pH resolution:
    0,01
  • Model:
    206 pH2
  • W×D×H:
    33×20×197 mm
  • Temperature range:
    0...+60 °C
  • Display:
    LCD, 2 lines
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Storage cap for testo 206, with KCI gel filling
    Storage cap for testo 206, with KCI gel filling
    TEST0554.2067
    Log in for availability
    1 * 1 ST - 20.20 SFr.

Product Information

  • Catalog #
    Availability
    Price
  • pH buffer solution 7,00 in dosing bottle (250 ml), with DAkkS calibration certificate
    pH buffer solution 7,00 in dosing bottle (250 ml), with DAkkS calibration certificate
    TEST0554.2063
    Log in for availability
    1 * 250 mL - 25.00 SFr.

Product Information

  • Catalog #
    Availability
    Price
  • pH buffer solution 4,01 in dosing bottle (250 ml), with DAkkS calibration certificate
    pH buffer solution 4,01 in dosing bottle (250 ml), with DAkkS calibration certificate
    TEST0554.2061
    Log in for availability
    1 * 250 mL - 25.00 SFr.

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.