Orders LinkContactUsLinkComponent
pH probes
pH probes
Spare pH probe for testo 206 pH2, with NTC, incl. gel storage cap, 0 - 14, 0...60 °C, Gel, 11×35 mm, Semi-solid substances
Catalog # TEST0650.2062
Supplier:   Testo
pH probes
Catalog # TEST0650.2062
Supplier:  Testo
pH probes
pH probes
Spare pH probe for testo 206 pH2, with NTC, incl. gel storage cap, 0 - 14, 0...60 °C, Gel, 11×35 mm, Semi-solid substances
Price
211.00 SFr.
Log in for availability
1 * 1 ST
Specifications
  • Ø×L:
    11×35 mm
  • For:
    Semi-solid substances
  • Description:
    Spare pH probe for testo 206 pH2, with NTC, incl. gel storage cap
  • Seq:
    03
  • Electrolyte:
    Gel
  • Temp.:
    0...60 °C
  • pH:
    0 - 14