Orders LinkContactUsLinkComponent
pH probes
pH probes
Spare pH probe for testo 206 pH1, with NTC, incl. gel storage cap, 0 - 14, 0...60 °C, Gel, 15×3,5 mm, Liquids
Catalog # TEST0650.2061
Supplier:   Testo
pH probes
Catalog # TEST0650.2061
Supplier:  Testo
pH probes
pH probes
Spare pH probe for testo 206 pH1, with NTC, incl. gel storage cap, 0 - 14, 0...60 °C, Gel, 15×3,5 mm, Liquids
Price
105.00 SFr.
Log in for availability
1 * 1 ST
Specifications
  • Ø×L:
    15×3,5 mm
  • For:
    Liquids
  • Description:
    Spare pH probe for testo 206 pH1, with NTC, incl. gel storage cap
  • Seq:
    02
  • Electrolyte:
    Gel
  • Temp.:
    0...60 °C
  • pH:
    0 - 14