Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Pyridine

Pyridine

Glass bottle, 500 mL
Catalog # ROTH9729.3
Flammable
Irritant
Marketsource
Supplier:   Roth Carl
Pyridine
Catalog # ROTH9729.3
Supplier:  Roth Carl
Marketsource
CAS Number:  110-86-1
Pyridine
Pyridine

Glass bottle, 500 mL

CAS Number:  110-86-1
Price
SFr. 98.30
Log in for availability
1 * 500 mL
Product Details & Documents
Synonyms:  Azabenzene, Azine
  • CAS Number:110-86-1
  • Formula:C₅H₅N
  • MDL Number:MFCD00011732
  • Molecular Weight:79.1 g/mol
  • Merck Index:13,08060
  • Boiling Pt:115 °C (1013 hPa)
  • Melting Pt:–44 °C
  • Density:0.983 g/cm³ (20 °C)
  • Flash Pt:17 °C
  • Storage Temperature:Ambient
  • EINECS:203-809-9
  • UN:1282
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.