Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Quartz wool

Quartz wool

Quartz wool
N° de catalogue ROTH9208.1
Marketsource
Fournisseur:   Roth Carl
Quartz wool
N° de catalogue ROTH9208.1
Fournisseur:  Roth Carl
Marketsource
Quartz wool

Quartz wool

Prix
SFr. 35.90
Connexion Pour la disponibilité
1 * 10 g
Product Details & Documents

This quartz wool is ideal for elementary analysis, as a final stopper in chromatography columns or as a filter aid.

  • Chemically pure
  • Fibre size 5 to 30 µm

Packaging: Packed in plastic bottles.

Spécifications
  • Description:
    Quartz wool
  • Seq:
    01