Tween® 20 (Polysorbate)
Glass bottle for solids, 250 g
ROTH9127.1
Marketsource
: Polyoxyethylene sorbitan monolaurate
- :9005-64-5
- :C₅₈H₁₁₄O₂₆
- :MFCD00165986
- :1227.53 g/mol
- :1.1 g/cm³ (20 °C)
- :Ambient
- :500-018-3
- Pack type:Glass bottle for solids
- Pk:250 g