Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Ethanol absolute

Ethanol absolute

25 L
Catalog # ROTH9065.6
Flammable
Ethanol absolute
Catalog # ROTH9065.6
Supplier:  
CAS Number:  64-17-5
Ethanol absolute

25 L

CAS Number:  64-17-5
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Ethanol absolute for analysis EMSURE® ACS,ISO,Reag. Ph Eur 1 * 25 L Catalog #1.00983.6025

Product 1.00983.6025 is similar to ROTH9065.6 but manufacturer and specifications may vary.

Ethanol absolute AnalaR NORMAPUR ACS/REAG.PE/REAG.USP 25L 1 * 25 L Catalog #20821.467

Product 20821.467 is similar to ROTH9065.6 but manufacturer and specifications may vary.

Product Details & Documents
Synonyms:  Ethyl alcohol
  • CAS Number:64-17-5
  • Formula:C₂H₅OH
  • MDL Number:MFCD00003568
  • Molecular Weight:46.07 g/mol
  • Merck Index:12,03806
  • Boiling Pt:78.3 °C (1013 hPa)
  • Melting Pt:–117 °C
  • Density:0.7895 g/cm³ (20 °C)
  • Flash Pt:12 °C
  • Storage Temperature:Ambient
  • EINECS:200-578-6
  • UN:1170
  • ADR:3,II
  • REACH:01-2119457610-43
Specifications

Specifications

  • Pk:
    25 L

Specifications