Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Caproic acid | Avantor

Caproic acid

Glass bottle, 100 mL
Katalognr. ROTH8799.1
Ätzend
Marketsource
Anbieter:   Roth Carl
Caproic acid
Katalognr. ROTH8799.1
Anbieter:  Roth Carl
Marketsource
CAS Nummer:  142-62-1
Caproic acid

Glass bottle, 100 mL

CAS Nummer:  142-62-1
Preis
SFr. 25.10
Einloggen für Verfügbarkeit
1 * 100 mL
Product Details & Documents
Synonyme:  Hexanoic acid
  • CAS Nummer:142-62-1
  • Formel:C₆H₁₂O₂
  • MDL-Nummer:MFCD00004421
  • Molekulargewicht:116.16 g/mol
  • Merck-Index:13,01765
  • Siedepunkt:202 °C (1013 hPa)
  • Schmelzender Platingehalt:–4.8 °C
  • Dichte:0.9276 g/cm³ (20 °C)
  • Blitz Pt:102 °C
  • Lagertemperatur:Ambient
  • EINECS:205-550-7
  • UN:2829
  • ADR:8,III
Technische Daten

Technische Daten

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Technische Daten

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.