Methanol
Glass bottle, 2,5 L
Catalog # ROTHX948.2
Marketsource
Supplier: Roth Carl
Methanol
Catalog # ROTHX948.2
Supplier: Roth Carl
Marketsource
CAS Number: 67-56-1
Jump to:
Product Details & Documents
Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
- CAS Number:67-56-1
- Formula:CH₃OH
- MDL Number:MFCD00004595
- Molecular Weight:32.04 g/mol
- Merck Index:13,05984
- Boiling Pt:64.6 °C (1013 hPa)
- Melting Pt:–98 °C
- Density:0.7918 g/cm³ (20 °C)
- Flash Pt:11 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:200-659-6
- UN:1230
- ADR:3,II
- REACH:01-2119433307-44
Specifications
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Cat. No.:ROTHX948.2
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Environmentally Preferable:Safer Human and Environmental Health
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



