Order Entry
Switzerland
Orders LinkContactUsLinkComponent
L(-)-Malic acid

L(-)-Malic acid

Glass bottle for solids, 10 g
Catalog # ROTH8684.1
Irritant
Marketsource
Supplier:   Roth Carl
L(-)-Malic acid
Catalog # ROTH8684.1
Supplier:  Roth Carl
Marketsource
CAS Number:  97-67-6
L(-)-Malic acid

Glass bottle for solids, 10 g

CAS Number:  97-67-6
Price
SFr. 34.70
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  L(-)-Malic acid, (S)-(-)-Hydroxy-1,4-butanedioic acid, (S)-(-)-Hydroxysuccinic acid, (S)-2-hydroxysuccinic acid, L(-)-Hydroxysuccinic acid, L(-)-Hydroxy-1,4-butanedioic acid, L-(–)-Malic Acid
  • CAS Number:97-67-6
  • Formula:C₄H₆O₅
  • MDL Number:MFCD00064213
  • Molecular Weight:134.09 g/mol
  • Merck Index:13,05730
  • Boiling Pt:322 °C (1013 hPa)
  • Melting Pt:101…104 °C
  • Density:1.595 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:202-601-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications