Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Sodium nitrite

Sodium nitrite

5 kg
Catalog # ROTH8604.2
Oxidizing
Toxic
Environment
Marketsource
Supplier:   Roth Carl
Sodium nitrite
Catalog # ROTH8604.2
Supplier:  Roth Carl
Marketsource
CAS Number:  7632-00-0
Sodium nitrite

5 kg

CAS Number:  7632-00-0
Price
SFr. 95.90
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  Diazotizing salts
  • CAS Number:7632-00-0
  • Formula:NaNO₂
  • MDL Number:MFCD00011118
  • Molecular Weight:69 g/mol
  • Merck Index:13,08721
  • Boiling Pt:320 °C (1013 hPa)
  • Melting Pt:284 °C
  • Density:2.168 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-555-9
  • UN:1500
  • ADR:5.1,III
Specifications

Specifications

  • Pk:
    5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.