Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Sodium nitrite

Sodium nitrite

Glass bottle for solids, 1 kg
Catalog # ROTH8604.1
Oxidizing
Toxic
Environment
Marketsource
Supplier:   Roth Carl
Sodium nitrite
Catalog # ROTH8604.1
Supplier:  Roth Carl
Marketsource
CAS Number:  7632-00-0
Sodium nitrite

Glass bottle for solids, 1 kg

CAS Number:  7632-00-0
Price
SFr. 24.60
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Diazotizing salts
  • CAS Number:7632-00-0
  • Formula:NaNO₂
  • MDL Number:MFCD00011118
  • Molecular Weight:69 g/mol
  • Merck Index:13,08721
  • Boiling Pt:320 °C (1013 hPa)
  • Melting Pt:284 °C
  • Density:2.168 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-555-9
  • UN:1500
  • ADR:5.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications