Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Dichloromethane

Dichloromethane

Glass bottle, 2,5 L
Catalog # ROTH8424.2
Health
Irritant
Marketsource
Supplier:   Roth Carl
Dichloromethane
Catalog # ROTH8424.2
Supplier:  Roth Carl
Marketsource
CAS Number:  75-09-2
Dichloromethane
Dichloromethane

Glass bottle, 2,5 L

CAS Number:  75-09-2
Price
SFr. 71.40
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Methylene chloride, DCM
  • CAS Number:75-09-2
  • Formula:CH₂Cl₂
  • MDL Number:MFCD00000881
  • Molecular Weight:84.93 g/mol
  • Merck Index:12,06140
  • Boiling Pt:39.8 °C (1013 hPa)
  • Melting Pt:–95 °C
  • Density:1.322 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-838-9
  • UN:1593
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications