Methanol
10 L
Katalognr. ROTH8388.3
Marketsource
Anbieter: Roth Carl
Methanol
Katalognr. ROTH8388.3
Anbieter: Roth Carl
Marketsource
CAS Nummer: 67-56-1
Direkt zu
Product Details & Documents
Synonyme: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
- CAS Nummer:67-56-1
- Formel:CH₃OH
- MDL-Nummer:MFCD00004595
- Molekulargewicht:32.04 g/mol
- Merck-Index:13,05984
- Siedepunkt:64.6 °C (1013 hPa)
- Schmelzender Platingehalt:–98 °C
- Dichte:0.7918 g/cm³ (20 °C)
- Blitz Pt:11 °C (closed cup)
- Lagertemperatur:Ambient
- EINECS:200-659-6
- UN:1230
- ADR:3,II
- ERREICHEN:01-2119433307-44
Technische Daten
Technische Daten
- Pk:10 L
- Cat. No.:ROTH8388.3
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Environmentally Preferable:Safer Human and Environmental Health
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



