Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Potassium nitrate | Avantor

Potassium nitrate

Glass bottle for solids, 2,5 kg
Catalog # ROTH8001.2
Oxidizing
Marketsource
Supplier:   Roth Carl
Potassium nitrate
Catalog # ROTH8001.2
Supplier:  Roth Carl
Marketsource
CAS Number:  7757-79-1
Potassium nitrate

Glass bottle for solids, 2,5 kg

CAS Number:  7757-79-1
Price
SFr. 63.00
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  Nitrate of Potash
  • CAS Number:7757-79-1
  • Formula:KNO₃
  • MDL Number:MFCD00011409
  • Molecular Weight:101.1 g/mol
  • Merck Index:13,07733
  • Melting Pt:334 °C
  • Density:2.109 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-818-8
  • UN:1486
  • ADR:5.1,III
  • REACH:01-2119488224-35

Dokumente

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2,5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.