Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Potassium nitrate

Potassium nitrate

Glass bottle for solids, 500 g
Catalog # ROTH8001.1
Oxidizing
Marketsource
Supplier:   Roth Carl
Potassium nitrate
Catalog # ROTH8001.1
Supplier:  Roth Carl
Marketsource
CAS Number:  7757-79-1
Potassium nitrate

Glass bottle for solids, 500 g

CAS Number:  7757-79-1
Price
22.20 SFr.
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Nitrate of Potash
  • CAS Number:7757-79-1
  • Formula:KNO₃
  • MDL Number:MFCD00011409
  • Molecular Weight:101.1 g/mol
  • Merck Index:13,07733
  • Melting Pt:334 °C
  • Density:2.109 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-818-8
  • UN:1486
  • ADR:5.1,III
  • REACH:01-2119488224-35

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications