Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Phenol

Glass bottle, 500 mL
Catalog # ROTH0038.3
Toxic
Corrosive
Health
Marketsource
Supplier:   Roth Carl
Phenol
Catalog # ROTH0038.3
Supplier:  Roth Carl
Marketsource
CAS Number:  108-95-2
Phenol

Glass bottle, 500 mL

CAS Number:  108-95-2
Price
115.10 SFr.
Log in for availability
1 * 500 mL
Product Details & Documents
Synonyms:  Phenylalcohol, Carbolic acid, Hydroxybenzene
  • CAS Number:108-95-2
  • Formula:C₆H₅OH
  • MDL Number:MFCD00002143
  • Molecular Weight:94.11 g/mol
  • Merck Index:12,07390
  • Boiling Pt:181.7 °C (1013 hPa)
  • Melting Pt:40.8 °C
  • Density:1.0722 g/cm³ (20 °C)
  • Flash Pt:81 °C
  • Storage Temperature:Ambient
  • EINECS:203-632-7
  • UN:1671
  • ADR:6.1,II
  • REACH:01-2119471329-32
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.