Formaldehyde 37% in aqueous solution
30 L
Katalognr. ROTHP733.3
Marketsource
Anbieter: Roth Carl
Formaldehyde 37% in aqueous solution
Katalognr. ROTHP733.3
Anbieter: Roth Carl
Marketsource
CAS Nummer: 50-00-0
Direkt zu
Product Details & Documents
Synonyme: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
- CAS Nummer:50-00-0
- Formel:CH₂O
- MDL-Nummer:MFCD00003274
- Molekulargewicht:30.03 g/mol
- Lagertemperatur:Ambient
- EINECS:200-001-8
- UN:2209
- ADR:8,III
Technische Daten
Technische Daten
- Pk:30 L



