Sodium nitrate
Glass bottle for solids, 1 kg
N° de catalogue ROTHA136.2
Marketsource
Fournisseur: Roth Carl
Aller à
Product Details & Documents
- Numéro CAS:7631-99-4
- Formule:NaNO₃
- Numéro MDL:MFCD00011119
- Poids moléculaire:84.99 g/mol
- Index Merck:13,08720
- Point d'ébullition:380 °C (1013 hPa)
- Point de fusion:308 °C
- Densité:2.26 g/cm³ (20 °C)
- Température de stockage:Ambient
- EINECS:231-554-3
- UN:1498
- ADR:5.1,III
- REACH:01-2119488221-41
Spécifications
Spécifications
- Pack type:Glass bottle for solids
- Pk:1 kg
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



