Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Urea

Urea

5 kg
Catalog # ROTH7638.2
Marketsource
Supplier:   Roth Carl
Urea
Catalog # ROTH7638.2
Supplier:  Roth Carl
Marketsource
CAS Number:  57-13-6
Urea
CAS Number:  57-13-6
Price
SFr. 123.00
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
  • CAS Number:57-13-6
  • Formula:NH₂CONH₂
  • MDL Number:MFCD00008022
  • Molecular Weight:60.06 g/mol
  • Merck Index:13,09935
  • Boiling Pt:197 °C (1013 hPa)
  • Melting Pt:132.7 °C
  • Density:1.34 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-315-5
  • REACH:01-2119463277-33
Specifications

Specifications

  • Pk:
    5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.