Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Oleic acid

Oleic acid

Glass bottle, 50 mL
Catalog # ROTH7213.2
Marketsource
Supplier:   Roth Carl
Oleic acid
Catalog # ROTH7213.2
Supplier:  Roth Carl
Marketsource
CAS Number:  112-80-1
Oleic acid
Oleic acid

Glass bottle, 50 mL

CAS Number:  112-80-1
Price
342.00 SFr.
Log in for availability
1 * 50 mL
Product Details & Documents
Synonyms:  (9Z)-9-Octadecenoic acid
  • CAS Number:112-80-1
  • Formula:C₁₈H₃₄O₂
  • MDL Number:MFCD00064242
  • Boiling Pt:286 °C (1013 hPa)
  • Density:0.8946 g/cm³ (20 °C)
  • Storage Temperature:Freezer
  • EINECS:204-007-1
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.