Dextrin, yellow
5 kg
Catalog # ROTH6777.2
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Dextrin
- CAS Number:9004-53-9
- Formula:(C₆H₁₂O₆)n
- MDL Number:MFCD00081554
- Molecular Weight:180.16 g/mol
- Storage Temperature:Ambient
- EINECS:232-675-4
Specifications
Specifications
- Pk:5 kg



